Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIVE/DEAD™ FungaLight™ Yeast Viability Kit, for flow cytometry

Catalog No. L34952
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 Assays kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L34952 200 Assays kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L34952 Supplier Invitrogen™ Supplier No. L34952
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The LIVE/DEAD FungaLight Yeast Viability Kit enables researchers to easily, reliably, and quantitatively distinguish live and dead yeast in minutes by flow cytometry.

The LIVE/DEAD™ FungaLight™ Yeast Viability Kit enables researchers to easily, reliably and quantitatively distinguish live and dead yeast in minutes by flow cytometry. The kit contains solutions of the SYTO 9 green-fluorescent nucleic acid stain and the red-fluorescent nucleic acid stain, propidium iodide. These stains differ both in their spectral characteristics and in their ability to penetrate healthy yeast cells. When used alone, the SYTO 9 stain generally labels all yeast in a population - those with intact membranes and those with damaged membranes. In contrast, propidium iodide penetrates only yeast with damaged membranes, causing a reduction in the SYTO 9 stain fluorescence by fluorescence resonance energy transfer (FRET) when both dyes are present. As a result, yeast with intact membranes stain fluorescent green, whereas yeast with damaged membranes stain fluorescent red.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Impermeant, Permeant
Color Green, Red
Content And Storage Contains 1 vial of SYTO™ 9 dye (solution in DMSO), and 1 vial of propidium iodide, 300 μL (solution in DMSO).

Store in freezer (-5°C to -30°C) and protect from light.
Description LIVE/DEAD™ FungaLight™ Yeast Viability Kit, for flow cytometry
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Yeast Viability Kit
Dye Type SYTO™ 9, propidium iodide
Form Solution
Format Tube(s)
Product Line FungaLight, LIVE/DEAD
Quantity 200 Assays kit
Shipping Condition Room Temperature
Excitation Wavelength 485/535 nm
Emission Wavelength 498/617
Show More Show Less
What are the fluorescence excitation/emission maxima for the SYTO 9 dye and propidium iodide in the LIVE/DEAD FungaLight Yeast Viability Kit, for flow cytometry?

SYTO 9 dye: 480/500 nm
Propidium iodide: 490/635 nm

How do I prepare dead cell controls for LIVE/DEAD cell viability assays?

There are two easy options. One is to heat-inactivate the cells by placing at 60 degrees C for 20 minutes. The second is to subject the cells to 70% ethanol. Alcohol-fixed cells can be stored indefinitely in the freezer until use, potentially up to several years.

Centrifuge cells, pellet, and remove supernatant.
Fix cells: Add 10 mL ice cold 70% ETOH to a 15 mL tube containing the cell pellet, adding dropwise at first while vortexing, mix well.
Store in freezer until use.
When ready to use, wash twice and resuspend in buffer of choice.

Can I look at bacteria and yeast on a flow cytometer?

Yes, you can. We offer:

-Counting assays: Bacteria Counting Kit, for flow cytometry (Cat. No. B7277) or LIVE/DEAD BacLight Bacterial Viability and Counting Kit, for flow cytometry (Cat. No. L34856).
-Viability/vitality assays: LIVE/DEAD BacLight Bacterial Viability Kit (Cat. No. L13152).
-Membrane potential: BacLight Bacterial Membrane Potential Kit (Cat. No. B34950).
-Yeast viability/vitality assays: LIVE/DEAD FungaLight Yeast Viability Kit, for flow cytometry (Cat. No. L34952), FungaLight Yeast CFDA, AM/Propidium Iodide Vitality Kit (Cat. No. F34953)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.