Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Livin Antibody, DyLight 350 , Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297074UV
Description
Livin Polyclonal antibody specifically detects Livin in Human samples. It is validated for ELISASpecifications
Livin | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
DyLight 350 | |
baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7 | |
Produced in rabbits immunized with purified, recombinant Human Livin (AAH14475.1; Met1-Ser298). | |
0.1 mL | |
Apoptosis, Zinc Finger | |
79444 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction