Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Livin Antibody (OTI1D12), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272206PECY7
Description
Livin Monoclonal specifically detects Livin in Human samples. It is validated for Flow Cytometry.Specifications
Livin | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
BIRC7 | |
Full length recombinant protein of human Livin (NP_071444) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
OTI1D12 | |
Flow Cytometry | |
baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7 | |
Mouse | |
Affinity Purified | |
Apoptosis, Zinc Finger | |
79444.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction