Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Livin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522330
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522330 Supplier Invitrogen™ Supplier No. PA522330
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The inhibitor of apoptosis (IAP) family of proteins regulates programmed cell death triggered by various stimuli. All IAPs have at least one baculovirus IAP repeat (BIR) motif that is essential for their anti-apoptotic activity. Recently, it has been identified that a novel human inhibitor of apoptosis protein (IAP) family member termed Livin or ML-IAP, which contains a single baculoviral IAP repeat (BIR) domain and a COOH-terminal RING finger domain. Livin gene has two splicing variants that contain open reading frames of 298 and 280 amino acids and both contained a single copy of baculovirus IAP repeat (BIR) and RING domain. Both of the isoforms inhibit TNFa induced apoptosis in Jurkat cells. Livin expression is low in adult tissues. It is relatively expressed at a higher level in developmental tissues and in many cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Livin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene BIRC7
Gene Accession No. Q96CA5
Gene Alias baculoviral IAP repeat containing 7; baculoviral IAP repeat-containing 7; baculoviral IAP repeat-containing 7 (livin); baculoviral IAP repeat-containing protein 7; Baculoviral IAP repeat-containing protein 7 30 kDa subunit; Baculoviral IAP repeat-containing protein 7 30kDa subunit; Birc7; E130019N06; KIAP; Kidney inhibitor of apoptosis protein; LIVIN; livin inhibitor of apoptosis; melanoma inhibitor of apoptosis protein; MLIAP; ML-IAP; RGD1562883; RING finger protein 50; RING-type E3 ubiquitin transferase BIRC7; RNF50; RP11-261N11.7; Truncated livin; UNQ5800/PRO19607/PRO21344
Gene Symbols BIRC7
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Livin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79444
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.