Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Livin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575440
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575440 Supplier Thermo Scientific Supplier No. PA575440
Add to Cart
Add to Cart

Livin Polyclonal Antibody

Livin Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Livin
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Gene BIRC7
Gene Accession No. A2AWP0, Q96CA5
Gene Alias baculoviral IAP repeat containing 7; baculoviral IAP repeat-containing 7; baculoviral IAP repeat-containing 7 (livin); baculoviral IAP repeat-containing protein 7; Baculoviral IAP repeat-containing protein 7 30 kDa subunit; Baculoviral IAP repeat-containing protein 7 30kDa subunit; Birc7; E130019N06; KIAP; Kidney inhibitor of apoptosis protein; LIVIN; livin inhibitor of apoptosis; melanoma inhibitor of apoptosis protein; MLIAP; ML-IAP; RGD1562883; RING finger protein 50; RING-type E3 ubiquitin transferase BIRC7; RNF50; RP11-261N11.7; Truncated livin; UNQ5800/PRO19607/PRO21344
Gene Symbols BIRC7
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 180-230 of Human Livin.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 329581, 79444
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.