Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIX1 Antibody (RM0282-4F47), DyLight 680, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211779FR
Description
LIX1 Monoclonal specifically detects LIX1 in Mouse samples. It is validated for Western Blot.Specifications
LIX1 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LIX1 | |
Mouse recombinant LIX1 | |
0.1 mL | |
167410 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0282-4F47 | |
Western Blot | |
C5orf11, chromosome 5 open reading frame 11, FLJ25534, limb expression 1, Lix1 homolog (chicken), Lix1 homolog (mouse), protein limb expression 1 homolog | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction