Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LIX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563571
Description
Immunogen sequence: RRITKEFIME SVQEAVASTS GTLDDADDPS TSVGAYHYML ESNMGKT Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
The specific function of this protein remains unknown.
Specifications
LIX1 | |
Polyclonal | |
Unconjugated | |
LIX1 | |
5730466L18Rik; C5orf11; Lft; limb and CNS expressed 1; limb expression 1; limb expression 1 homolog; limb expression 1 homolog (chicken); Lix1; Lix1 homolog; Lix1 homolog (chicken); Lowfat homolog; protein limb expression 1 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
167410 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N485 | |
LIX1 | |
Recombinant protein corresponding to Human LIX1. Recombinant protein control fragment (Product #RP-101817). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction