Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113153
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113153 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113153 Supplier Invitrogen™ Supplier No. PA5113153
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LIX1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The specific function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIX1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LIX1
Gene Accession No. Q6P566, Q8N485
Gene Alias 5730466L18Rik; C5orf11; Lft; limb and CNS expressed 1; limb expression 1; limb expression 1 homolog; limb expression 1 homolog (chicken); Lix1; Lix1 homolog; Lix1 homolog (chicken); Lowfat homolog; protein limb expression 1 homolog
Gene Symbols LIX1
Host Species Rabbit
Immunogen Recombinant Human Protein limb expression 1 homolog protein (1-282AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 167410, 66643
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.