Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIX1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566604
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566604 Supplier Invitrogen™ Supplier No. PA566604
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAVLREAVEAV VRSFAKHTQG YGRVNVVEAL QEFWQMKQSR GADLKNGALV VYEMVPSNSP PYV Highest antigen sequence identity to the following orthologs - mouse 100%, rat 98%.

LIX1L is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIX1L
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LIX1L
Gene Accession No. Q8IVB5
Gene Alias D130027M04Rik; limb and CNS expressed 1 like; Lix1 homolog (chicken) like; Lix1 homolog-like; LIX1L; Lix1-like; LIX1-like protein
Gene Symbols LIX1L
Host Species Rabbit
Immunogen Recombinant Human LIX1L. Recombinant protein control fragment (Product #RP-107259).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128077
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.