Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LKAAEAR1 Antibody (Z36P2C1*E7), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250477G
Description
LKAAEAR1 Monoclonal specifically detects LKAAEAR1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
C20orf201 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LKAAEAR1 | |
Peptide Sequence - AIRLELFLPP (immunogen peptide is identical in mouse, rat, sheep, bovine and equine) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
Z36P2C1*E7 | |
Western Blot, ELISA, Immunohistochemistry | |
bA299N6.3, chromosome 20 open reading frame 201, hypothetical protein LOC198437, LKAAEAR motif containing 1, LKAAEAR1 | |
Mouse | |
Protein A purified | |
RUO | |
198437 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction