Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LKAAEAR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563307
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563307 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563307 Supplier Invitrogen™ Supplier No. PA563307
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDRQERRRVE TILEENVDGT IFPR Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 92%.

LKAAEAR1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LKAAEAR1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LKAAEAR1
Gene Accession No. Q8TD35
Gene Alias bA299N6.3; C20orf201; LKAAEAR motif containing 1; LKAAEAR motif-containing protein 1; LKAAEAR1; Protein LKAAEAR1
Gene Symbols LKAAEAR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LKAAEAR1. Recombinant protein control fragment (Product #RP-101675).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 198437
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.