Learn More
Invitrogen™ LKB1 Monoclonal Antibody (B.593.0)

Description
It is not recommended to aliquot this antibody.
Specifications
Specifications
| Antigen | LKB1 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | B.593.0 |
| Concentration | 45 μg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 |
| Gene | STK11 |
| Gene Accession No. | Q15831 |
| Gene Alias | AA408040; CAMK group; CAMKL family; hLKB1; liver kinase B1; Liver kinase B1 homolog; LKB 1; LKB1; LKB-1; LKB1(S); Par-4; PJS; polarization-related protein LKB1; R75140; renal carcinoma antigen NY-REN-19; serine/threonine kinase 11; serine/threonine-protein kinase 11; serine/threonine-protein kinase LKB1; serine/threonine-protein kinase STK11; STK11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.