Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LKB1/STK11 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP214835AF532
Description
LKB1/STK11 Polyclonal specifically detects LKB1/STK11 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LKB1/STK11 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
EC 2.7.11.1, LKB1 serine/threonine kinase 11 (Peutz-Jeghers syndrome), PJS, PJS polarization-related protein LKB1, Renal carcinoma antigen NY-REN-19, serine/threonine kinase 11, serine/threonine-protein kinase 11, Serine/threonine-protein kinase LKB1, STK11 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
STK11 | |
A synthetic peptide made to a internal portion of the human LKB1 protein (between residues 250-350) [UniProt Q15831] | |
0.1 mL | |
Hypoxia Signaling, mTOR Pathway, p53 Pathway, Phospho Specific, Protein Kinase | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction