Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LKB1/STK11 Antibody (OTI0A3), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271300IR
Description
LKB1/STK11 Monoclonal specifically detects LKB1/STK11 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LKB1/STK11 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
STK11 | |
Recombinant protein fragment corresponding to amino acids 1-236 of human LKB1/STK11 (NP_000446) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI0A3 | |
Western Blot, Immunohistochemistry | |
EC 2.7.11.1, LKB1 serine/threonine kinase 11 (Peutz-Jeghers syndrome), PJS, PJS polarization-related protein LKB1, Renal carcinoma antigen NY-REN-19, serine/threonine kinase 11, serine/threonine-protein kinase 11, Serine/threonine-protein kinase LKB1, STK11 | |
Mouse | |
Affinity Purified | |
Hypoxia Signaling, mTOR Pathway, p53 Pathway, Phospho Specific, Protein Kinase | |
6794.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction