Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LL-37 Antibody (B-T37), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318120IR
Description
LL-37 Monoclonal antibody specifically detects LL-37 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
LL-37 | |
Monoclonal | |
DyLight 755 | |
18 kDa cationic antimicrobial protein, Antibacterial peptide FALL-39, CAP18, CAP-18, cathelicidin antimicrobial peptide, CRAMP, epididymis secretory sperm binding protein, FALL39, FALL-39, FALL-39 peptide antibiotic, hCAP-181, HSD26, LL37 | |
Human antimicrobial peptide LL-37. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
0.1 mL | |
Innate Immunity | |
820 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, ELISA | |
B-T37 | |
50mM Sodium Borate | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction