Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LL-37 Mouse anti-Human, Clone: B-F57, Novus Biologicals™

Mouse Monoclonal Antibody
$366.50 - $699.00
Specifications
Antigen | LL-37 |
---|---|
Clone | B-F57 |
Concentration | 1 mg/ml |
Dilution | Western Blot, ELISA |
Applications | Western Blot, ELISA |
Description
LL-37 Monoclonal antibody specifically detects LL-37 in Human samples. It is validated for Western Blot, ELISASpecifications
LL-37 | |
1 mg/ml | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Innate Immunity | |
PBS. Sterile-filtered through 0.22 um. | |
820 | |
IgG1 κ | |
Affinity purified |
B-F57 | |
Western Blot, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
18 kDa cationic antimicrobial protein, Antibacterial peptide FALL-39, CAP18, CAP-18, cathelicidin antimicrobial peptide, CRAMP, epididymis secretory sperm binding protein, FALL39, FALL-39, FALL-39 peptide antibiotic, hCAP-181, HSD26, LL37 | |
Human antimicrobial peptide LL-37. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
Primary | |
Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title