Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LL-37 Mouse anti-Human, Clone: B-L47, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318121500UL
This item is not returnable.
View return policy
Description
LL-37 Monoclonal antibody specifically detects LL-37 in Human samples. It is validated for ELISASpecifications
| LL-37 | |
| Monoclonal | |
| 1 mg/ml | |
| ELISA | |
| 18 kDa cationic antimicrobial protein, Antibacterial peptide FALL-39, CAP18, CAP-18, cathelicidin antimicrobial peptide, CRAMP, epididymis secretory sperm binding protein, FALL39, FALL-39, FALL-39 peptide antibiotic, hCAP-181, HSD26, LL37 | |
| Human antimicrobial peptide LL-37. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
| 500 μg | |
| Innate Immunity | |
| 820 | |
| Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| ELISA | |
| B-L47 | |
| Unconjugated | |
| PBS. Sterile-filtered through 0.22 um. | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction