Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LLGL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120697
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120697 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120697 Supplier Invitrogen™ Supplier No. PA5120697
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: OVCAR3, A-431, U-251MG, Mouse testis, Mouse brain, Rat testis. Immunogen sequence: MMKFRFRRQG ADPQREKLKQ ELFAFNKTVE HGFPNQPSAL AFDPELRIMA IGTRSGAVKI YGAPGVEFTG LHRDAATVTQ MHFLTGQGRL LSLLDDSSLH LWEIVHHNGC AHLEEALSFQ LPSRPGFDGA SAPLSLTRVT VVLLVAAGDI AALGTEGSSV FFLDVTTLTL LEGQTLAPGE VLRSVPDDYR.

LLGL1 is a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene for LLGL1 is located within the Smith-Magenis syndrome region on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LLGL1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene LLGL1
Gene Accession No. Q15334, Q80Y17, Q8K4K5
Gene Alias AI325176; DLG4; HUGL; Hugl protein; HUGL1; Hugl-1; Human homolog to the D-lgl gene protein; lethal giant larvae homolog 1; lethal giant larvae homolog 1 (Drosophila); lethal giant larvae homolog 1, scribble cell polarity complex component; lethal(2) giant larvae protein homolog 1; Lgl1; LLGL; Llgl1; LLGL1, scribble cell polarity complex component; Llglh; Mgl1; Mgl-1; Mlgl; Rgl1; Rgl-1
Gene Symbols LLGL1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16897, 3996, 54265
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.