Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LLGL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576364
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
LLGL1 is a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene for LLGL1 is located within the Smith-Magenis syndrome region on chromosome 17. LLGL2 is a protein similar to lethal (2) giant larvae of Drosophila. In fly, the protein's ability to localize cell fate determinants is regulated by the atypical protein kinase C (aPKC). In human, this protein interacts with aPKC-containing complexes and is cortically localized in mitotic cells.
Specifications
LLGL2 | |
Polyclonal | |
Unconjugated | |
LLGL2 | |
9130006H11Rik; HGL; Hugl-2; human giant larvae homolog; lethal giant larvae homolog 2; lethal giant larvae homolog 2 (Drosophila); lethal giant larvae homolog 2, scribble cell polarity complex component; lethal giant larvae-like protein 2; Lethal(2) giant larvae protein homolog 2; LGL2; LLGL scribble cell polarity complex component 2; Llgl2; LLGL2, scribble cell polarity complex component; Llglh2; RGD1560307 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
217325, 360661, 3993 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3TJ91, Q6P1M3 | |
LLGL2 | |
Recombinant fusion protein of mouse LLGL2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction