Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LLGL2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986159
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6159 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6159 Supplier Bioss Supplier No. BS4771R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LLGL1 is a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene for LLGL1 is located within the Smith-Magenis syndrome region on chromosome 17. LLGL2 is a protein similar to lethal (2) giant larvae of Drosophila. In fly, the protein′s ability to localize cell fate determinants is regulated by the atypical protein kinase C (aPKC). In human, this protein interacts with aPKC-containing complexes and is cortically localized in mitotic cells.

Specifications

Antigen LLGL2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LLGL2
Gene Accession No. Q6P1M3
Gene Alias 9130006H11Rik; HGL; Hugl-2; human giant larvae homolog; lethal giant larvae homolog 2; lethal giant larvae homolog 2 (Drosophila); lethal giant larvae homolog 2, scribble cell polarity complex component; Lethal giant larvae-like protein 2; Lethal(2) giant larvae protein homolog 2; LGL2; LLGL scribble cell polarity complex component 2; Llgl2; LLGL2, scribble cell polarity complex component; Llglh2; RGD1560307
Gene Symbols LLGL2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LLGL2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3993
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.