Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LLPH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566207
Description
Immunogen sequence: RSKWKRKMRAE KRKKNAPKEA SRLKSILKLD GDVLMKDVQE IATVVVPKPK Highest antigen sequence identity to the following orthologs - mouse 82%, rat 84%.
In hippocampal neurons, regulates dendritic and spine growth and synaptic transmission.
Specifications
LLPH | |
Polyclonal | |
Unconjugated | |
LLPH | |
C12orf31; cPERP-G; hLLP; human LAPS18-like protein; LLP homolog, long-term synaptic facilitation; LLP homolog, long-term synaptic facilitation (Aplysia); LLPH; Protein LAPS18-like; Protein LLP homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84298 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRT6 | |
LLPH | |
Recombinant Human LLPH. Recombinant protein control fragment (Product #RP-105113). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction