Learn More
Tocris Bioscience™ LM11A 31 dihydrochloride
Description
Specifications
Specifications
| Purity | 95% |
| Molecular Formula | (2 S,3 S)-2-Amino-3-methyl-N-[2-(4-morpholinyl)ethyl]pentanamide dihydrochloride |
| CAS | 1243259-19-9 |
| Content And Storage | Desiccate at RT |
| Quantity | 10 mg |
| Product Type | LM11A 31 dihydrochloride |
| Molecular Weight (g/mol) | M.W. (Observed): 316.27 |
| Inhibitors | LM11A 31 dihydrochloride |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.