Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LM11A 31 dihydrochloride
SDP

Catalog No. 504650R Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-465-0R 50 mg
50-461-0R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-465-0R Supplier Tocris Bioscience™ Supplier No. 5046/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nonpeptide p75NTR ligand; neuroprotective

LM11A 31 dihydrochloride is a nonpeptide p75NTR ligand and nerve growth factor (NGF) antagonist. LM11A 31 blocks p75-mediated cell death, inhibits proNGF induced cell death (IC50 = 1-10 nM), increases proliferation and survival of hippocampal neural progenitors and inhibits Aβ-induced cell death (EC50 = 20 nM). In animal models of Alzheimer's disease LM11A 31 prevents and reverses atrophy of cholinergic neurites and reverses Alzheimer's pathology; in models of Huntington's disease LM11A 31 increases survival, reduces the formation of aggregates of mutant huntingtin and neuronal degeneration. Also promotes functional recovery in a mouse model of spinal cord injury. Orally available and brain penetrant.

Specifications

Purity 95%
Molecular Formula (2 S,3 S)-2-Amino-3-methyl-N-[2-(4-morpholinyl)ethyl]pentanamide dihydrochloride
CAS 1243259-19-9
Content And Storage Desiccate at RT
Quantity 50 mg
Product Type LM11A 31 dihydrochloride
Molecular Weight (g/mol) M.W. (Observed): 316.27
Inhibitors LM11A 31 dihydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.