Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LM22B 10
GREENER_CHOICESDP

Catalog No. 603750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-375-0 50 mg
60-371-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-375-0 Supplier Tocris Bioscience™ Supplier No. 6037/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

TrkB and TrkC agonist; brain penetrant

LM22B 10 is a TrkB and TrkC agonist. Exhibits neurotrophic activity (EC50 = 200-300 nM), improves survival and increases neurite outgrowth in hippocampal cells in vitro. Exhibits no significant activity in a screen of 57 other receptors and transporters. Inhibits binding of BDNF and neurotrophin-3 to TrkB and TrkC, respectively. Active in vivo and brain penetrant.

Specifications

Chemical Name or Material 2,2′,2′,2′′′-[[(4-Chlorophenyl)methylene]bis(4,1-phenylenenitrilo)]tetrakisethanol
CAS 342777-54-2
Quantity 50 mg
Molecular Formula C27H33ClN2O4
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.