Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMAN1 Monoclonal Antibody (OTI1B8), TrueMAB™, OriGene
SDP

Catalog No. 502541669
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1669 30 μL
50-167-3338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1669 Supplier OriGene Supplier No. TA502269S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LMAN1 Monoclonal antibody specifically detects LMAN1 in Human samples. It is validated for Western Blot

Specifications

Antigen LMAN1
Applications Western Blot
Classification Monoclonal
Clone OTI1B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CTDSP1
Gene Accession No. Q9GZU7
Gene Alias 2610020P13Rik; AI326273; AU043785; C730041J05; EGK_09725; endoplasmic reticulum-golgi intermediate compartment protein 53; ERGIC53; ERGIC-53; ER-Golgi intermediate compartment 53 kDa protein; F5F8D; FMFD1; Gp58; intracellular mannose-specific lectin MR60; Lectin mannose-binding 1; lectin, mannose binding 1; lectin, mannose-binding, 1; LMAN1; MCFD1; MR60; P58; Protein ERGIC-53
Host Species Mouse
Immunogen Full length human recombinant protein of human CTDSP1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58190
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.