Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMAN1 Mouse anti-Human, Clone: OTI1C5, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA502237

Description
The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.Specifications
LMAN1 | |
Monoclonal | |
0.97 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P49257 | |
LMAN1 | |
Full length human recombit protein of human LMAN1 produced in Hek293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1C5 | |
Unconjugated | |
LMAN1 | |
ERGIC-53, ERGIC53, F5F8D, FMFD1, MCFD1, MR60, gp58 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3998 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction