Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMAN1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA1074

Catalog No. PA1074

  • $516.65 / Each of 1

    Save 21%

    Reg : $655.50

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Edge
Add to Cart

Description

Description

PA1-074 detects ERGIC-53 in rat, mouse and human samples. PA1-074 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects a ~57.5 kDa protein representing ERGIC-53 in rat brain samples. The PA1-074 immunogen is a synthetic peptide corresponding to residues F(159) D S F D N D G K K N N P A I(173) of rat ERGIC-53. This sequence is conserved in mouse and human. The PA1-074 immunizing peptide (Cat. # PEP-277) is available for use in neutralization and control experiments.

The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LMAN1
Polyclonal
Unconjugated
LMAN1
2610020P13Rik; AI326273; AU043785; C730041J05; EGK_09725; endoplasmic reticulum-golgi intermediate compartment protein 53; ERGIC53; ERGIC-53; ER-Golgi intermediate compartment 53 kDa protein; F5F8D; FMFD1; Gp58; intracellular mannose-specific lectin MR60; Lectin mannose-binding 1; lectin, mannose binding 1; lectin, mannose-binding, 1; LMAN1; MCFD1; MR60; P58; Protein ERGIC-53
Rabbit
Antigen affinity chromatography
RUO
116666, 3998, 70361
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P49257, Q62902, Q9D0F3
LMAN1
Synthetic peptide corresponding to residues F(159) D S F D N D G K K N N P A I(173) of rat ERGIC-53.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.