Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMAN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587900
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587900 Supplier Invitrogen™ Supplier No. PA587900
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGKEPPTPDK EISEKEKEKY QEEFEHFQQE LDKKKEEFQK GHPDLQGQPA EEIFESVGDR ELRQVFEGQN RIHLEIKQLN RQLDMILDEQ RRYVSSLTEE ISKRGAGMPG QHGQITQQEL DTVVKTQHEI LRQVNEMKNS MSETVRLVSG MQHPGSAGGV YETTQHFIDI KEHLHIVKRD IDNLVQRNMP SNEKPKCPEL PPFPSCLSTV H; Positive Samples: BT-474, LO2, U-251MG; Cellular Location: Endoplasmic reticulum membrane, Endoplasmic reticulum-Golgi intermediate compartment membrane, Golgi apparatus membrane, Single-pass membrane protein, Single-pass type I membrane protein.

The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMAN1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LMAN1
Gene Accession No. P49257, Q62902, Q9D0F3
Gene Alias 2610020P13Rik; AI326273; AU043785; C730041J05; EGK_09725; endoplasmic reticulum-golgi intermediate compartment protein 53; ERGIC53; ERGIC-53; ER-Golgi intermediate compartment 53 kDa protein; F5F8D; FMFD1; Gp58; intracellular mannose-specific lectin MR60; Lectin mannose-binding 1; lectin, mannose binding 1; lectin, mannose-binding, 1; LMAN1; MCFD1; MR60; P58; Protein ERGIC-53
Gene Symbols LMAN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 270-480 of human LMAN1 (NP_0055611).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116666, 3998, 70361
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.