Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMAN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | LMAN1 |
---|---|
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
LMAN1 Polyclonal specifically detects LMAN1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LMAN1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
3998 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
ERGIC-53, ERGIC53ER-Golgi intermediate compartment 53 kDa protein, F5F8Dendoplasmic reticulum-golgi intermediate compartment protein 53, FMFD1, Gp58, intracellular mannose specific lectin, Intracellular mannose-specific lectin MR60, Lectin mannose-binding 1, lectin, mannose-binding, 1, MCFD1coagulation factor V-factor VIII combined deficiency, MR60, protein ERGIC-53 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 270-480 of human LMAN1 (NP_005561.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title