Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lman1l Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547805
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547805 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547805 Supplier Invitrogen™ Supplier No. PA547805
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Lman1l
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene LMAN1L
Gene Accession No. Q5FB95
Gene Alias complexin III; ERGIC-53L; ERGIC-53-like protein; ERGIC53-like protein; Ergl; Lectin mannose-binding 1-like; lectin, mannose binding 1 like; lectin, mannose-binding, 1 like; Lman1l; LMAN1-like protein; Protein ERGIC-53-like; slamp; Sublingual acinar membrane protein; UNQ2784/PRO7174
Gene Symbols LMAN1L
Host Species Sheep
Immunogen E. coli-derived recombinant rat LMAN1L/SLAMP (aa 33-436).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300743
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.