Learn More
Invitrogen™ LMAN1L Polyclonal Antibody

Description
LMAN1L Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | LMAN1L |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | LMAN1L |
| Gene Accession No. | Q9HAT1 |
| Gene Alias | complexin III; ERGIC-53L; ERGIC-53-like protein; ERGIC53-like protein; Ergl; Lectin mannose-binding 1-like; lectin, mannose binding 1 like; lectin, mannose-binding, 1 like; Lman1l; LMAN1-like protein; Protein ERGIC-53-like; slamp; Sublingual acinar membrane protein; UNQ2784/PRO7174 |
| Gene Symbols | LMAN1L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.