Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMAN2 Antibody (02), DyLight 350 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305830UV
Description
LMAN2 Monoclonal antibody specifically detects LMAN2 in Human samples. It is validated for Western Blot, ELISA, ELISA, ELISASpecifications
LMAN2 | |
Monoclonal | |
DyLight 350 | |
C5orf8, chromosome 5 open reading frame 8, Glycoprotein GP36b, GP36B, Lectin mannose-binding 2, lectin, mannose-binding 2, vesicular integral protein of 36 kDa, vesicular integral-membrane protein VIP36, VIP36 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LMAN2 (Q12907; Asp 45-Arg 322). | |
0.1 mL | |
Signal Transduction | |
10960 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
02 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction