Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMAN2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116337
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116337 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116337 Supplier Invitrogen™ Supplier No. PA5116337
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LMAN2L Polyclonal Antibody for Western Blot

May be involved in the regulation of export from the endoplasmic reticulum of a subset of glycoproteins. May function as a regulator of ERGIC-53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMAN2L
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene LMAN2L
Gene Accession No. Q9H0V9
Gene Alias A630028F14Rik; Lectin mannose-binding 2-like; lectin, mannose binding 2 like; lectin, mannose-binding 2-like; LMAN2L; LMAN2-like protein; MRT52; PSEC0028; UNQ368/PRO704; VIP36-like; VIP36-like protein; VIPL
Gene Symbols LMAN2L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 322-348 amino acids from the C-terminal region of human LMAN2L.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81562
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.