Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMBR1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | LMBR1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
LMBR1 Polyclonal specifically detects LMBR1 in Human samples. It is validated for Western Blot.Specifications
LMBR1 | |
Polyclonal | |
Rabbit | |
Q8WVP7 | |
64327 | |
Synthetic peptides corresponding to LMBR1(limb region 1 homolog (mouse)) The peptide sequence was selected from the N terminal of LMBR1. Peptide sequence MEGQDEVSAREQHFHSQVRESTICFLLFAILYVVSYFIITRYKRKSDEQE. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
ACHPchromosome 7 open reading frame 2, C7orf2FLJ11665, DIF14, Differentiation-related gene 14 protein, limb region 1 homolog (mouse), limb region 1 protein homolog, PPD2, TPT | |
LMBR1 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title