Learn More
Invitrogen™ LMBR1 Monoclonal Antibody (4A1)

Description
Peptide Sequence: SRMFTVMGQL LVKPTILEDL DEQIYIITLE EEALQRRLNG LSSSVEYNIM ELEQELENVK TLKTKLERRK KASAWERNLV YP.
Specifications
Specifications
| Antigen | LMBR1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4A1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Lmbr1 |
| Gene Accession No. | Q8WVP7 |
| Gene Alias | 1110048D14Rik; ACHP; AU017641; C7orf2; DIF14; Differentiation-related gene 14 protein; FLJ11665; limb development membrane protein 1; limb region 1; limb region 1 homolog; limb region 1 protein; Limb region 1 protein homolog; Lmbr1; LSS; PPD2; THYP; TPT; Unknown (protein for MGC:134350); ZRS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.