Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMBR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543411
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543411 Supplier Invitrogen™ Supplier No. PA543411
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MEGQDEVSAR EQHFHSQVRE STICFLLFAI LYVVSYFIIT RYKRKSDEQE Sequence homology: Cow: 100%; Dog: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

This gene encodes a member of the LMBR1-like membrane protein family. Another member of this protein family has been shown to be a lipocalin transmembrane receptor. A highly conserved, cis-acting regulatory module for the sonic hedgehog gene is located within an intron of this gene. Consequently, disruption of this genic region can alter sonic hedgehog expression and affect limb patterning, but it is not known if this gene functions directly in limb development. Mutations and chromosomal deletions and rearrangements in this genic region are associated with acheiropody and preaxial polydactyly, which likely result from altered sonic hedgehog expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMBR1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Lmbr1
Gene Accession No. Q8WVP7
Gene Alias 1110048D14Rik; ACHP; AU017641; C7orf2; DIF14; Differentiation-related gene 14 protein; FLJ11665; limb development membrane protein 1; limb region 1; limb region 1 homolog; limb region 1 protein; Limb region 1 protein homolog; Lmbr1; LSS; PPD2; THYP; TPT; Unknown (protein for MGC:134350); ZRS
Gene Symbols Lmbr1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LMBR1 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64327
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.