Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMBR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553452
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553452 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553452 Supplier Invitrogen™ Supplier No. PA553452
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FTVMGQLLVK PTILEDLDEQ IYIITLEEEA LQRRLNGLSS SVEYNIMELE QELENVKTLK TKLERRKKAS AWERN Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

This gene encodes a member of the LMBR1-like membrane protein family. Another member of this protein family has been shown to be a lipocalin transmembrane receptor. A highly conserved, cis-acting regulatory module for the sonic hedgehog gene is located within an intron of this gene. Consequently, disruption of this genic region can alter sonic hedgehog expression and affect limb patterning, but it is not known if this gene functions directly in limb development. Mutations and chromosomal deletions and rearrangements in this genic region are associated with acheiropody and preaxial polydactyly, which likely result from altered sonic hedgehog expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMBR1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Lmbr1
Gene Accession No. Q8WVP7
Gene Alias 1110048D14Rik; ACHP; AU017641; C7orf2; DIF14; Differentiation-related gene 14 protein; FLJ11665; limb development membrane protein 1; limb region 1; limb region 1 homolog; limb region 1 protein; Limb region 1 protein homolog; Lmbr1; LSS; PPD2; THYP; TPT; Unknown (protein for MGC:134350); ZRS
Gene Symbols Lmbr1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LMBR1. Recombinant protein control fragment (Product #RP-91225).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64327
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.