Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMBR1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526795
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526795 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526795 Supplier Invitrogen™ Supplier No. PA526795
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LMBR1L Polyclonal Antibody for Western Blot, IHC (P), Flow

LMBR1L, also known as LIMR (lipocalin-1-interacting membrane receptor), interacts with lipocalin-1 (Lcn-1), a lipocalin member produced by a number of secretory glands and tissues and known to bind a variety of lipophilic compounds. LMBR1L is a 489-amino acid protein with a predicted molecular mass of 55 kDa and 9 putative transmembrane helices. LMBR1L has been shown to mediate cellular uptake of LMBR1L, thus it is characterized as a endocytic receptor. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMBR1L
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene LMBR1L
Gene Accession No. Q6UX01
Gene Alias 1110013E13Rik; AA415653; D15Ertd735e; KIAA1174; limb development membrane protein 1 like; limb development membrane protein 1-like; limb region 1 homolog-like; limb region 1 like; limb region 1 protein homolog-like; limb region 1-like homolog; limb region 1-like protein -like; LIMR; lipocalin-1 interacting membrane receptor; lipocalin-1-interacting membrane receptor; lipocalin-interacting membrane receptor; Lmbr1l; Protein LMBR1L; RGD1309631; UNQ458/PRO783; Uteroglobin receptor
Gene Symbols LMBR1L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 462-489 amino acids from the C-terminal region of human LMBR1L.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55716
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Is the FOXG1 Polyclonal Antibody (Cat. No. PA526794) validated for immunocytochemistry/immunofluorescence (ICC/IF) applications?

Yes, the FOXG1 Polyclonal Antibody (Cat. No. PA526794) has been validated for ICC/IF applications.

WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.