Learn More
Invitrogen™ LMBRD1 Polyclonal Antibody

Description
Immunogen sequence: KGTRSAAYER LENTEDIEEV EQHIQTIKSK SKDGRPLPAR DKRALKQFEE RLRTLKKRER HLEFIENSWW TKFCGALRP Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 91%.
Specifications
Specifications
| Antigen | LMBRD1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Lmbrd1 |
| Gene Accession No. | Q9NUN5 |
| Gene Alias | 0910001K20Rik; AV347960; BM-021; C6orf209; CD001; HDAg-L-interacting protein NESI; hepatitis delta antigen-L interacting protein; liver regeneration p-53 related protein; LMBD1; LMBR1 domain containing 1; LMBR1 domain-containing protein 1; LMBRD1; MAHCF; MSTP044; NESI; nuclear export signal-interacting protein; Probable lysosomal cobalamin transporter; protein N90b |
| Gene Symbols | Lmbrd1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.