Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMCD1 Antibody (OTI1H5), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271865MFV500
Description
LMCD1 Monoclonal antibody specifically detects LMCD1 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
LMCD1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
29995 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI1H5 | |
Flow Cytometry | |
dyxin, LIM and cysteine-rich domains 1, LIM and cysteine-rich domains protein 1 | |
Full length human recombinant protein of human LMCD1(NP_055398) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction