Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMCD1 Antibody (OTI1H5), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271865PEATT594
Description
LMCD1 Monoclonal specifically detects LMCD1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
LMCD1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
LMCD1 | |
Full length recombinant protein of human LMCD1 (NP_055398) produced in HEK293T cells. | |
0.1 mL | |
29995.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI1H5 | |
Flow Cytometry | |
dyxin, LIM and cysteine-rich domains 1, LIM and cysteine-rich domains protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction