Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LMCD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549414
Description
LMCD1 Polyclonal Antibody for Western Blot
LMCD1 encodes a member of the LIM-domain family of zinc finger proteins. The encoded protein contains an N-terminal cysteine-rich domain and two C-terminal LIM domains. The presence of LIM domains suggests involvement in protein-protein interactions. The protein may act as a co-regulator of transcription along with other transcription factors. Alternate splicing results in multiple transcript variants of this gene.
Specifications
LMCD1 | |
Polyclonal | |
Unconjugated | |
LMCD1 | |
AW455500; AW553074; dyxin; LIM and cysteine rich domains 1; LIM and cysteine-rich domains 1; LIM and cysteine-rich domains protein 1; LIMD1; LMCD1 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
29995, 30937, 494021 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8VEE1, Q9NZU5 | |
LMCD1 | |
KLH conjugated synthetic peptide between 316-350 amino acids from the C-terminal region of human LMCD1. | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction