Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMCD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554969
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554969 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554969 Supplier Invitrogen™ Supplier No. PA554969
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLPIYDQDPS RCRGLLENEL KLMEEFVKQY KSEALGVGEV ALPGQGGLPK EEGKQQEKPE GAETTAATTN GSLSDPSKEV EYVCELCKGA APPDSPVVYS DRAGYNKQWH PTCFVCAKCS EPLVDLIYFW KDGA Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

LMCD1 encodes a member of the LIM-domain family of zinc finger proteins. The encoded protein contains an N-terminal cysteine-rich domain and two C-terminal LIM domains. The presence of LIM domains suggests involvement in protein-protein interactions. The protein may act as a co-regulator of transcription along with other transcription factors. Alternate splicing results in multiple transcript variants of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMCD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LMCD1
Gene Accession No. Q9NZU5
Gene Alias AW455500; AW553074; dyxin; LIM and cysteine rich domains 1; LIM and cysteine-rich domains 1; LIM and cysteine-rich domains protein 1; LIMD1; LMCD1
Gene Symbols LMCD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LMCD1. Recombinant protein control fragment (Product #RP-100927).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29995
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.