Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543420 Supplier Invitrogen™ Supplier No. PA543420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MRPDSPTMAA PAESLRRRKT GYSDPEPESP PAPGRGPAGS PAHLHTGTFW Sequence homology: Human: 100%.

Involved in the maturation of specific proteins in the endoplasmic reticulum. Required for maturation and transport of active lipoprotein lipase (LPL) through the secretory pathway. Each LMF1 molecule chaperones 50 or more molecules of LPL. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMF1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LMF1
Gene Accession No. Q96S06
Gene Alias 2400010G15Rik; AW822050; C16orf26; cld; FLJ12681; FLJ22302; HMFN1876; JFP11; Lipase maturation factor 1; LMF1; RGD1310180; Tmem112; TMEM112A; transmembrane protein 112
Gene Symbols LMF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LMF1 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64788
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.