Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LMF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564041
Description
Immunogen sequence: YVEPGTHGRL WTGAHRLFGA VEHLQLANSY GLFRRMTGLG GRPEVVLEGS YDGHHWTEIE FMY Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Involved in the maturation of specific proteins in the endoplasmic reticulum. May be required for maturation and transport of active lipoprotein lipase (LPL) through the secretory pathway.
Specifications
LMF2 | |
Polyclonal | |
Unconjugated | |
LMF2 | |
AI451006; Lipase maturation factor 2; LMF2; RGD1306274; TMEM112B; Tmem153; Transmembrane protein 112B; transmembrane protein 153 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91289 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BU23 | |
LMF2 | |
Recombinant protein corresponding to Human LMF2. Recombinant protein control fragment (Product #RP-103459). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction