Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597930
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597930 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597930 Supplier Invitrogen™ Supplier No. PA597930
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LMF2 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Involved in the maturation of specific proteins in the endoplasmic reticulum. May be required for maturation and transport of active lipoprotein lipase (LPL) through the secretory pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMF2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LMF2
Gene Accession No. Q8C3X8, Q9BU23
Gene Alias AI451006; Lipase maturation factor 2; LMF2; RGD1306274; TMEM112B; Tmem153; Transmembrane protein 112B; transmembrane protein 153
Gene Symbols LMF2
Host Species Rabbit
Immunogen Recombinant Human Lipase maturation factor 2 protein (432-636AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105847, 91289
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.