Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMF2 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. FEPA5145467
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145467 200 μL
FEPA5145468 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145467 Supplier Invitrogen™ Supplier No. PA5145467
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Involved in the maturation of specific proteins in the endoplasmic reticulum. May be required for maturation and transport of active lipoprotein lipase (LPL) through the secretory pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMF2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene LMF2
Gene Accession No. Q9BU23
Gene Alias AI451006; Lipase maturation factor 2; LMF2; RGD1306274; TMEM112B; Tmem153; Transmembrane protein 112B; transmembrane protein 153
Gene Symbols LMF2
Host Species Rabbit
Immunogen Recombinant Human LMF2 protein, Leu454-Lys707 (Accession #Q9BU23).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100518488, 91289
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.