Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 162491AP150UL

Description
Involved in the maturation of specific proteins in the endoplasmic reticulum. May be required for maturation and transport of active lipoprotein lipase (LPL) through the secretory pathway.Specifications
LMF2 | |
Polyclonal | |
Unconjugated | |
LMF2 | |
lipase maturation factor 2, LMF2, TMEM112B, TMEM153, Transmembrane protein 112B, Transmembrane protein 153 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
105847, 315218, 91289 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A1L1J9, Q8C3X8, Q9BU23 | |
Lmf2 | |
LMF2 Fusion Protein Ag9208 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction