Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30445920UL
Description
LMO1 Polyclonal specifically detects LMO1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
LMO1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
Cysteine-rich protein TTG-1, LIM domain only 1 (rhombotin 1), LMO-1, RBTN1MGC116692, RHOM1LIM domain only protein 1, rhombotin-1, TTG1T-cell translocation protein 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-156 of human LMO1 (NP_002306.1). | |
20 μL | |
Cancer | |
4004 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction