Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMO2 Antibody (LMO2/3147R), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308945JF646
Description
LMO2 Monoclonal antibody specifically detects LMO2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ ImmunofluorescenceSpecifications
LMO2 | |
Monoclonal | |
Janelia Fluor 646 | |
Cysteine-rich protein TTG-2, LIM domain only 2 (rhombotin-like 1), LIM domain only protein 2, LMO-2, RBTN2rhombotin-2, RBTNL1rhombotin-like 1, RHOM2T-cell translocation protein 2, T-cell translocation gene 2, TTG2rhombotin 2 | |
Recombinant fragment (around aa 23-140) human LMO2 protein (exact sequence is proprietary) (Uniprot: P25791) | |
100 μg | |
Angiogenesis, Cancer | |
4005 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, Immunofluorescence | |
LMO2/3147R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction